Learn More
3-Penten-1-yne (Mixture of cis/trans), TRC
Product Code. 30178935
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178935 100 mg 100mg
30178919 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30178935 Supplier Toronto Research Chemicals Supplier No. TRCP227430100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2206-23-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.1
Synonym 2-Penten-4-yne, Propenylacetylene;
IUPAC Name (E)-pent-3-en-1-yne
SMILES CC(=O)N[C@@H](C)C(O)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C5H6/c1-3-5-4-2/h1,4-5H,2H3/b5-4+
Formula Weight 66.047
Chemical Name or Material 3-Penten-1-yne (Mixture of cis/trans)