Learn More
3-(Pentafluorothio)phenol, 97%, Thermo Scientific™
Product Code. 15456647
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456647 1 g 1g
15466647 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15456647 Supplier Thermo Scientific Alfa Aesar Supplier No. H33787.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 672-31-1
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788515
InChI Key KVOACEHJTNSNBD-UHFFFAOYSA-N
Synonym 3-pentafluorothio phenol, 3-pentafluorosulfanyl phenol, 3-pentafluoro-$l^ 6-sulfanyl phenol, 3-pentafluoro-??-sulfanyl phenol, 3-pentafluorosulfanylphenol, 3-pentafluorosulfur phenol, 3-hydroxyphenylpentafluorosulfur vi, 3-pentafluoro-lambda-sulfanyl phenol, 3-pentafluoro-lambda∼6∼-sulfanyl phenol, 3-pentakis fluoranyl-$l^ 6-sulfanyl phenol
PubChem CID 2779201
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC(=CC=C1)S(F)(F)(F)(F)F
Specifications
Quantity 1 g
Formula Weight 220.16
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)phenol