missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzotrifluoride, 97%

Product Code. p-7044343
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312306 25 g 25g
11322306 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312306 Supplier Thermo Scientific Alfa Aesar Supplier No. L05948.14
Add to basket
This item is not returnable. View return policy

CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-46-4
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007260
InChI Key WHNAMGUAXHGCHH-UHFFFAOYSA-N
Synonym 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
PubChem CID 7386
IUPAC Name 1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point -5°C
Boiling Point 201°C to 205°C
Flash Point 87°C (188°F)
Odor Characteristic
Quantity 25 g
UN Number UN2306
Beilstein 2051362
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzotrifluoride
compliance-icons
Product Identifier
  • 3-Nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: m-Nitrobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.