Learn More
3-Nitrobenzoic acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 3-Nitrobenzoic acid |
| CAS | 121-92-6 |
| Density | 1.49 g/cm³ at 20°C |
| Flash Point | 189.6°C (Pensky-Martens closed cup) |
| Melting Range | 139°C to 142°C |
| Molecular Formula | 3-(NO2)C6H4COOH |
| Quantity | 25 kg |
| Synonym | 3-Nitrobenzoic acid, 3-Nitrobenzoic acid |
| Molecular Weight (g/mol) | 167.12 |
| Grade | Not Available |