missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonic Acid Sodium Salt, Sigma-Aldrich™

Product Code. 15884191
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884191 1 kg 1kg
15874191 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884191 Supplier Sigma Aldrich Supplier No. 8065481000
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-68-4
Grade Research
Molecular Formula 3-(NO2)C6H4SO3Na
Molecular Weight (g/mol) 225.15

Specifications

Melting Point 350°C (Decomposition)
Density 0.45 g/cm3 at 20°C
MDL Number MFCD00007490
Quantity 1 kg
Synonym 3-Nitrobenzenesulfonic acid sodium salt, Sodium 3-nitrobenzenesulfonate, 3-Nitrobenzenesulfonic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.