Learn More
3-Nitro Ropivacaine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Nitro Ropivacaine |
| CAS | 247061-07-0 |
| Molecular Formula | C17H25N3O3 |
| Quantity | 500 mg |
| Synonym | (2S)-N-(2,6-Dimethyl-3-nitrophenyl)-1-propyl-2-piperidinecarboxamide |
| InChI Formula | InChI=1S/C17H25N3O3/c1-4-10-19-11-6-5-7-15(19)17(21)18-16-12(2)8-9-14(13(16)3)20(22)23/h8-9,15H,4-7,10-11H2,1-3H3,(H,18,21)/t15-/m0/s1 |
| SMILES | CCCN1CCCC[C@H]1C(=O)Nc2c(C)ccc(c2C)[N+](=O)[O-] |
| IUPAC Name | (2S)-N-(2,6-dimethyl-3-nitrophenyl)-1-propylpiperidine-2-carboxamide |
| Molecular Weight (g/mol) | 319.4 |
| Formula Weight | 319.1896 |