Description
- Designed for smooth operation and long-lasting performance
- This spare part guarantees precise control and consistent results
- Restore the precision of your Eppendorf mechanical pipette with an original replacement for a worn or damaged piston
Specifications
Specifications
| For Use With | Pipettes |
| Material | Thermoplastic |
| Type | Piston |
| Volume (Metric) | 2.5 μL |
| For Use With (Equipment) | Pipette Tips |
| Quantity | 1 Pk. |
| Warranty | 10 μL |
| Width (English) | 200 μL |
| Width (Metric) | 300 μL |
| Weight (English) | 20 μL |
| Show More |