missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)phenylacetic acid, 95%, Thermo Scientific™

Product Code. 11357309
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357309 1 g 1g
11367309 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357309 Supplier Thermo Scientific Alfa Aesar Supplier No. H27452.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18698-73-2
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.24
MDL Number MFCD03701554
InChI Key AKYCOAMUNLESIH-UHFFFAOYSA-N
Synonym 3-methylthio phenylacetic acid, 2-3-methylthio phenyl acetic acid, 3-methylthio phenyl acetic acid, benzeneacetic acid, 3-methylthio, 3-methylsulfanyl phenyl acetic acid, acmc-20anie, 2-3-methylsulfanylphenyl acetic acid, 3-carboxymethyl thioanisole, 3-methylthiophenylacetic acid, 3-methylthiobenzeneacetic acid
PubChem CID 2737461
IUPAC Name 2-(3-methylsulfanylphenyl)acetic acid
SMILES CSC1=CC=CC(CC(O)=O)=C1

Specifications

Melting Point 77°C to 81°C
Quantity 1 g
Formula Weight 182.24
Percent Purity 95%
Chemical Name or Material 3-(Methylthio)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.