Learn More
3-(Methylthio)butanal, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Methylthio)butanal |
| CAS | 16630-52-7 |
| Molecular Formula | C5H10OS |
| Quantity | 50 g |
| Synonym | 3-(Methylthio)butyraldehyde, 3-Methylsulfanylbutyraldehyde, beta-(Methylthio)butyraldehyde |
| InChI Formula | InChI=1S/C5H10OS/c1-5(7-2)3-4-6/h4-5H,3H2,1-2H3 |
| SMILES | CSC(C)CC=O |
| IUPAC Name | 3-methylsulfanylbutanal |
| Molecular Weight (g/mol) | 118.2 |
| Formula Weight | 118.0452 |