Learn More
3-Methylglutarylcarnitine Chloride, TRC
Product Code. 17993805
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993805 25 mg 25mg
17922845 50 mg 50mg
17953545 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993805 Supplier Toronto Research Chemicals Supplier No. TRCM22641525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 821794-55-2
Molecular Formula C13 H24 N O6 . Cl
Molecular Weight (g/mol) 325.78
Synonym 3-carboxy-2-(4-carboxy-3-methyl-1-oxobutoxy)-N,N,N-trimethyl-1-propanaminium Chloride
IUPAC Name [3-carboxy-2-(4-carboxy-3-methyl-butanoyl)oxy-propyl]-trimethyl-ammonium;chloride
SMILES [Cl-].CC(CC(=O)O)CC(=O)OC(CC(=O)O)C[N+](C)(C)C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C13H23NO6.ClH/c1-9(5-11(15)16)6-13(19)20-10(7-12(17)18)8-14(2,3)4;/h9-10H,5-8H2,1-4H3,(H-,15,16,17,18);1H
Formula Weight 325.1292
Chemical Name or Material 3-Methylglutarylcarnitine Chloride