Learn More
3-Methylbenzenesulfonamide, TRC
Product Code. 30483129
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483129 25 mg 25mg
30476861 50 mg 50mg
30466184 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30483129 Supplier Toronto Research Chemicals Supplier No. TRCM33655025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1899-94-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.22
IUPAC Name 3-methylbenzenesulfonamide
SMILES Cc1cccc(c1)S(=O)(=O)N
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C7H9NO2S/c1-6-3-2-4-7(5-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)
Formula Weight 171.0354
Chemical Name or Material 3-Methylbenzenesulfonamide