missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-hexanamine Hydrochloride Salt, TRC

Product Code. 17912275
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912275 50 mg 50mg
17920495 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17912275 Supplier Toronto Research Chemicals Supplier No. TRCM26368550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855475-52-4
Molecular Formula C7H17N . HCl
Molecular Weight (g/mol) 115.223646
Synonym 2-Amino-3-methylhexane Hydrochloride Salt
SMILES CC(C(N)C)CCC.Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H17N.ClH/c1-4-5-6(2)7(3)8;/h6-7H,4-5,8H2,1-3H3;1H
Formula Weight 151.1128
Chemical Name or Material 3-Methyl-2-hexanamine Hydrochloride Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.