Learn More
3-Methyl-2-butanethiol, 95%, remainder mainly 2-methyl-1-butanethiol, Thermo Scientific™
Product Code. 11486036
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486036 5 g 5g
11496036 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11486036 Supplier Thermo Scientific Alfa Aesar Supplier No. B22330.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2084-18-6
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00039648
InChI Key BFLXFRNPNMTTAA-UHFFFAOYNA-N
Synonym 3-methyl-2-butanethiol, 2-butanethiol, 3-methyl, 3-methyl-2-butylthiol, fema no. 3304, 3-methyl-2-butane thiol, acmc-209ff6, 3-methyl-2-butyl mercaptan, ksc202s8n, 1,2-dimethylpropyl hydrosulfide, 1,2-dimethylpropyl hydrosulfide #
PubChem CID 519823
IUPAC Name 3-methylbutane-2-thiol
SMILES CC(C)C(C)S
Tekniske data
Density 0.84
Boiling Point 100°C
Flash Point 3°C (37°F)
Odor Stench
Refractive Index 1.444
Quantity 5 g
UN Number UN3336
Sensitivity Air Sensitive
Formula Weight 104.22
Percent Purity 95%
Assay remainder mainly 2-methyl-1-butanethiol
Chemical Name or Material 3-Methyl-2-butanethiol
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.