Learn More
3-Methyl-1-pentanol, 98%
Product Code. 11426086
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426086 25 g 25g
11416086 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426086 Supplier Thermo Scientific Alfa Aesar Supplier No. B22356.14
Request Bulk or Custom Format
CAS: 589-35-5 | C6H14O | 102.177 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 589-35-5
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002937
InChI Key IWTBVKIGCDZRPL-UHFFFAOYSA-N
Synonym 3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
PubChem CID 11508
ChEBI CHEBI:87381
IUPAC Name 3-methylpentan-1-ol
SMILES CCC(C)CCO
Specifications
Density 0.823
Boiling Point 151°C to 152°C
Flash Point 58°C (136°F)
Refractive Index 1.418
Quantity 25 g
UN Number UN1987
Beilstein 1718979
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-pentanol