Learn More
3-Methoxy Favipiravir, TRC
Product Code. 30179457
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179457 5 mg 5mg
30179283 1 mg 1mg
30179320 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179457 Supplier Toronto Research Chemicals Supplier No. TRCM0033505MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 259794-09-7
Molecular Formula C6H6FN3O2
Molecular Weight (g/mol) 171.13
Synonym 6-Fluoro-3-methoxy-2-pyrazinecarboxamide;
IUPAC Name 6-fluoro-3-methoxypyrazine-2-carboxamide
SMILES COc1ncc(F)nc1C(=O)N
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C6H6FN3O2/c1-12-6-4(5(8)11)10-3(7)2-9-6/h2H,1H3,(H2,8,11)
Formula Weight 171.0444
Chemical Name or Material 3-Methoxy Favipiravir