Learn More
3-Methoxy-4-methylaniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 16452-01-0 |
| Molecular Formula | C8H11NO |
| MDL Number | MFCD00025371 |
| Quantity | 25 g |
| SMILES | C1=C(N)C=CC(=C1OC)C |
| Molecular Weight (g/mol) | 137.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Methoxy-4-methylaniline |