missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methoxybenzoic acid, 98%, Thermo Scientific™

Product Code. 11436526
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11436526

Brand: Thermo Scientific Alfa Aesar B24783.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68507-19-7
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00017347
InChI Key HNJSYSWRPCCSQI-UHFFFAOYSA-N
PubChem CID 624158
IUPAC Name 3-iodo-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)I

Specifications

Melting Point 238°C to 241°C
Quantity 25 g
Sensitivity Light sensitive
Synonym 3-iodo-4-methoxy-benzoic acid, 3-iodo-p-anisic acid, benzoic acid, 3-iodo-4-methoxy, 3-iodo-4-methoxybenzoicacid, pubchem4973, acmc-209o3r, akos bbb/204, benzoic acid,3-iodo-4-methoxy, 3-iodo-4-methoxybenzoic acid
Formula Weight 278.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methoxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.