Learn More
3-Iodo-1,2-benzenedicarboxylic Acid 1,2-Dimethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Iodo-1,2-benzenedicarboxylic Acid 1,2-Dimethyl Ester |
| CAS | 102928-38-1 |
| Molecular Formula | C10H9O4I |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H9IO4/c1-14-9(12)6-4-3-5-7(11)8(6)10(13)15-2/h3-5H,1-2H3 |
| SMILES | COC(=O)c1cccc(I)c1C(=O)OC |
| IUPAC Name | dimethyl 3-iodobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 320.08 |
| Formula Weight | 319.9546 |