missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl Phosphate, Disodium Salt, >98%, MP Biomedicals™

Product Code. 11407532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407532 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407532 Supplier MP Biomedicals Supplier No. 0215416783
  • ONLINE EXCLUSIVE PRICE
    221.00 EUR
    210.00 EUR / 250mg
    Save 11.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

3-Indoxyl Phosphate Disodium Salt is used as a histochemical substrate to demonstrate alkaline phosphatase

TRUSTED_SUSTAINABILITY

Specifications

CAS 3318-43-2
Color Gray
Assay Percent Range > 98%
Molecular Formula C8H6NNa2O4P
MDL Number MFCD00040646
Quantity 250 mg
Synonym 3-indoxyl phosphate disodium salt, disodium indol-3-yl phosphate, indol-3-yl sodium phosphate, disodium 1h-indol-3-yl phosphate, unii-p1w2d6403e, indoxylphosphate, disodium salt, sodium 1h-indol-3-yl phosphate, 1h-indol-3-ol, dihydrogen phosphate ester , disodium salt, ip, na2, 3-indoxyl phosphate
InChI Key WCSKWBKPKXJWEG-UHFFFAOYSA-L
SMILES C1=CC=C2C(=C1)C(=CN2)OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;1H-indol-3-yl phosphate
Molecular Weight (g/mol) 257.092
PubChem CID 76830
Formula Weight 257.1
Physical Form Powder
Chemical Name or Material 3-Indoxyl Phosphate, Disodium Salt, FTIR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.