missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionamide, TRC

Product Code. 30012676
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012676 50 mg 50mg
30003840 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012676 Supplier Toronto Research Chemicals Supplier No. TRCH95252550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2651-43-6
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
IUPAC Name 3-hydroxypropanamide
SMILES NC(=O)CCO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H7NO2/c4-3(6)1-2-5/h5H,1-2H2,(H2,4,6)
Formula Weight 89.0477
Chemical Name or Material 3-Hydroxypropionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.