Learn More
3-Hydroxyhexanoic Acid, TRC
Product Code. 30402955
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30402955 Supplier Toronto Research Chemicals Supplier No. TRCH8253201G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxyhexanoic Acid
CAS 10191-24-9
Purity Grade Notes GC
Molecular Formula C6 H12 O3
Quantity 1 g
Synonym (+/-)-3-Hydroxyhexanoic Acid, 3-Hydroxycaproic Acid, 3-Hydroxyhexanoic Acid, β-Hydroxy-n-caproic Acid, β-Hydroxycaproic Acid, β-Hydroxyhexanoic Acid
InChI Formula InChI=1S/C6H12O3/c1-2-3-5(7)4-6(8)9/h5,7H,2-4H2,1H3,(H,8,9)
SMILES CCCC(O)CC(=O)O
IUPAC Name 3-hydroxyhexanoic acid
Molecular Weight (g/mol) 132.158
Formula Weight 132.0786
Percent Purity >95