missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyhexanedioic Acid, TRC

Product Code. 30399241
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399241 5 mg
30394978 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399241 Supplier Toronto Research Chemicals Supplier No. TRCH9431005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14292-29-6
Molecular Formula C6 H10 O5
Molecular Weight (g/mol) 162.14
Synonym Hexanedioic acid, 3-hydroxy-, 3-Hydroxyhexanedioic acid, 3-Hydroxyadipic acid, β-Hydroxyadipic acid
IUPAC Name 3-hydroxyhexanedioic acid
SMILES OC(CCC(=O)O)CC(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H10O5/c7-4(3-6(10)11)1-2-5(8)9/h4,7H,1-3H2,(H,8,9)(H,10,11)
Formula Weight 162.0528
Chemical Name or Material 3-Hydroxyhexanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.