missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyglutaric Acid, TRC

Product Code. 30134494
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134494 5 mg 5mg
30007711 10 mg 10mg
30008223 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134494 Supplier Toronto Research Chemicals Supplier No. TRCH9425805MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-18-6
Molecular Formula C5 H8 O5
Molecular Weight (g/mol) 148.11
Synonym Pentanedioic acid, 3-hydroxy-, Glutaric acid, 3-hydroxy- (7CI,8CI), 3-Hydroxypentanedioic acid, 2,4-Dideoxypentaric acid, 3-Hydroxyglutaric acid, β-Hydroxyglutaric acid
IUPAC Name 3-hydroxypentanedioic acid
SMILES OC(CC(=O)O)CC(=O)O

Specifications

Quantity 5 mg
InChI Formula disodium;pentakis(azanylidynemethyl)-(oxoniumylidyne-λ^{4}-azanyl)iron(2+);dihydrate
Formula Weight 148.0372
Percent Purity >95%
Chemical Name or Material 3-Hydroxyglutaric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.