Learn More
3-Hydroxychrysene, TRC
Product Code. 30006609
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006609 100 mg 100mg
30004526 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006609 Supplier Toronto Research Chemicals Supplier No. TRCH750055100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63019-39-6
Molecular Formula C18 H12 O
Molecular Weight (g/mol) 244.29
Synonym 3-Chrysenol;
IUPAC Name chrysen-3-ol
SMILES Oc1ccc2ccc3c4ccccc4ccc3c2c1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C18H12O/c19-14-8-5-13-7-9-16-15-4-2-1-3-12(15)6-10-17(16)18(13)11-14/h1-11,19H
Formula Weight 244.0888
Chemical Name or Material 3-Hydroxychrysene