Learn More
3-Hydroxy-4-methoxycinnamic Acid-d3 (Isoferulic Acid-d3), TRC
Product Code. 30398125
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398125 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398125 Supplier Toronto Research Chemicals Supplier No. TRCH94618210MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic Acid-d3 (Isoferulic Acid-d3)
CAS 1028203-97-5
Molecular Formula C10H7D3O4
Quantity 10 mg
Synonym 3-[3-Hydroxy-4-(methoxy-d3)phenyl]acrylic Acid, Isoferulic Acid, 3-[3-Hydroxy-4-(methoxy-d3)phenyl]-2-propenoic Acid
InChI Formula InChI=1S/C10H10O4/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+/i1D3
SMILES OC1=C(OC([2H])([2H])[2H])C=CC(/C=C/C(O)=O)=C1
Molecular Weight (g/mol) 197.2
Formula Weight 197.0767