Learn More
3-Hydroxy-2-naphthohydrazide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 5341-58-2 |
| Molecular Formula | C11H10N2O2 |
| MDL Number | MFCD00004097 |
| Quantity | 25 g |
| SMILES | O=C(NN)C1=C(O)C=C2C=CC=CC2=C1 |
| Molecular Weight (g/mol) | 202.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Hydroxy-2-naphthohydrazide |