Learn More
3-Hexanone-2,2,4,4-d4, CDN
Product Code. 30315527
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315527 1 g 1g
30311054 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315527 Supplier CDN Supplier No. CDND80951G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hexanone-2,2,4,4-D4
CAS 24588-54-3
Molecular Formula C6 D4 H8 O
Quantity 1 g
Synonym 3-Hexanone-2,2,4,4-d4 (8 CI, 9 CI), 3-Hexanone-2,2,4,4-D4
InChI Formula InChI=1 S/C6H12O/c1-3-5-6(7)4-2/h3-5H2,1-2H3/i4D2,5D2
SMILES [2 H]C([2 H])(C)C(=O)C([2 H])([2 H])CC
IUPAC Name 2,2,4,4-tetradeuteriohexan-3-one
Molecular Weight (g/mol) 104.1835
Formula Weight 104.1139 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity