Learn More
3-Fluorocinnamic Acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 458-46-8 |
| Molecular Formula | C9H7FO2 |
| MDL Number | MFCD00004383 |
| Quantity | 5 g |
| SMILES | O=C(O)C=CC1=CC=CC(F)=C1 |
| Molecular Weight (g/mol) | 166.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Fluorocinnamic Acid |