Learn More
3-Fluorobenzyl chloride, 97%
Product Code. 11480533
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11480533
Brand: Thermo Scientific Alfa Aesar A12665.22
Request Bulk or Custom Format
CAS: 456-42-8 | C7H6ClF | 144.573 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 456-42-8
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000904
InChI Key XBDXMDVEZLOGMC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl chloride, 1-chloromethyl-3-fluorobenzene, m-fluorobenzyl chloride, benzene, 1-chloromethyl-3-fluoro, 3-fluorobenzylchloride, alpha-chloro-3-fluorotoluene, alpha-chloro-m-fluorotoluene, fluorobenzyl-3 chloride, toluene, .alpha.-chloro-m-fluoro, .alpha.-chloro-m-fluorotoluene
PubChem CID 9974
IUPAC Name 1-(chloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCl
Specifications
Density 1.213
Boiling Point 176°C to 177°C
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.512
Quantity 100 g
UN Number UN2920
Beilstein 742265
Formula Weight 144.58
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzyl chloride