CAS RN 456-42-8
CAS RN 456-42-8
IUPAC Name:
1-(chloromethyl)-3-fluorobenzene
Synonyms:
M-fluorobenzyl chloride
Molecular Weight (g/mol):
144.57
Molecular Formula:
C7H6ClF
InChi Key:
XBDXMDVEZLOGMC-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(CCl)=C1
1
–
1
of
1
results
1
3-Fluorobenzyl chloride, 97%
CAS: 456-42-8 Molecular Formula: C7H6ClF Molecular Weight (g/mol): 144.573 MDL Number: MFCD00000904 InChI Key: XBDXMDVEZLOGMC-UHFFFAOYSA-N Synonym: 3-fluorobenzyl chloride,1-chloromethyl-3-fluorobenzene,m-fluorobenzyl chloride,benzene, 1-chloromethyl-3-fluoro,3-fluorobenzylchloride,alpha-chloro-3-fluorotoluene,alpha-chloro-m-fluorotoluene,fluorobenzyl-3 chloride,toluene, .alpha.-chloro-m-fluoro,.alpha.-chloro-m-fluorotoluene PubChem CID: 9974 IUPAC Name: 1-(chloromethyl)-3-fluorobenzene SMILES: C1=CC(=CC(=C1)F)CCl