Learn More
3-Fluorobenzyl bromide, 95%
Product Code. 11472797
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472797 50 g 50g
11319489 5 g 5g
11329489 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11472797 Supplier Thermo Scientific Alfa Aesar Supplier No. A15446.18
Request Bulk or Custom Format
CAS: 456-41-7 | C7H6BrF | 189.027 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 456-41-7
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00000340
InChI Key SCBZBMXPJYMXRC-UHFFFAOYSA-N
Synonym 3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
PubChem CID 68007
IUPAC Name 1-(bromomethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CBr
Specifications
Density 1.546
Boiling Point 200°C
Flash Point 91°C (196°F)
Odor Pungent
Refractive Index 1.546
Quantity 50 g
UN Number UN3265
Beilstein 636503
Sensitivity Moisture sensitive
Formula Weight 189.03
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzyl bromide