Learn More
3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals
Product Code. p-7043589
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391287 100 mg 100mg
11381287 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11391287 Supplier Thermo Scientific Alfa Aesar Supplier No. L01479.MC
Add to basket
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
Solubility
Soluble in water (28 g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 403-90-7
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00063075
InChI Key VIIAUOZUUGXERI-UHFFFAOYNA-N
Synonym 3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine
PubChem CID 92100
ChEBI CHEBI:32771
IUPAC Name 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(F)=C1)C(O)=O
Specifications
Melting Point ∽280°C (decomposition)
Beilstein 3204804
Solubility Information Soluble in water (28g/L).
Quantity 100 mg
Formula Weight 199.18
Percent Purity 98%
Chemical Name or Material 3-Fluoro-DL-tyrosine