missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 11476186
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476186 1 g 1g
11486186 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476186 Supplier Thermo Scientific Alfa Aesar Supplier No. B23012.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 235106-09-9
Molecular Formula C8H7F4N
Molecular Weight (g/mol) 193.15
MDL Number MFCD00236287
InChI Key XYWYEOHNURYGGP-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzylamine, 3-fluoro-4-trifluoromethyl phenyl methanamine, 3-fluoro-4-trifluoromethyl phenyl methylamine, 1-3-fluoro-4-trifluoromethyl phenyl methanamine, acmc-1chwh, 3-fluoro-4-trifluoromethylbenzylamine, 3-fluoro-4-trifluoromethyl-benzylamine, benzenemethanamine,3-fluoro-4-trifluoromethyl
PubChem CID 2737567
IUPAC Name [3-fluoro-4-(trifluoromethyl)phenyl]methanamine
SMILES NCC1=CC=C(C(F)=C1)C(F)(F)F

Especificaciones

Refractive Index 1.453
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzylamine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.