missing translation for 'onlineSavingsMsg'
Learn More

3-[Ethyl(methyl)carbamoyl]benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15429885
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15429885

Brand: Thermo Scientific Alfa Aesar H52530.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871333-07-2
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.036
MDL Number MFCD08056369
InChI Key WVUBMQPFOZJFAA-UHFFFAOYSA-N
Synonym 3-ethyl methyl carbamoyl phenyl boronic acid, 3-ethyl methyl carbamoyl phenylboronic acid, 3-n-ethyl-n-methylamino carbonyl benzeneboronic acid, boronic acid,b-3-ethylmethylamino carbonyl phenyl, acmc-209qhz, 3-ethyl methyl carbamoyl benzeneboronic acid, 3-ethyl methyl carbamoyl phenylboronicacid, 3-ethyl methyl carbamoyl phenyl boronicacid
PubChem CID 44119833
IUPAC Name [3-[ethyl(methyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)N(C)CC)(O)O

Specifications

Melting Point 73°C to 76°C
Quantity 250 mg
Formula Weight 207.04
Percent Purity 98%
Chemical Name or Material 3-[Ethyl(methyl)carbamoyl]benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.