CAS RN 871333-07-2
CAS RN 871333-07-2
IUPAC Name:
{3-[ethyl(methyl)carbamoyl]phenyl}boronic acid
Synonyms:
3-[ethyl(methyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
207.04
Molecular Formula:
C10H14BNO3
InChi Key:
WVUBMQPFOZJFAA-UHFFFAOYSA-N
SMILES:
CCN(C)C(=O)C1=CC=CC(=C1)B(O)O
1
–
1
of
1
results
1
3-[Ethyl(methyl)carbamoyl]benzeneboronic acid, 98%, Thermo Scientific™
CAS: 871333-07-2 Molecular Formula: C10H14BNO3 Molecular Weight (g/mol): 207.036 MDL Number: MFCD08056369 InChI Key: WVUBMQPFOZJFAA-UHFFFAOYSA-N Synonym: 3-ethyl methyl carbamoyl phenyl boronic acid,3-ethyl methyl carbamoyl phenylboronic acid,3-n-ethyl-n-methylamino carbonyl benzeneboronic acid,boronic acid,b-3-ethylmethylamino carbonyl phenyl,acmc-209qhz,3-ethyl methyl carbamoyl benzeneboronic acid,3-ethyl methyl carbamoyl phenylboronicacid,3-ethyl methyl carbamoyl phenyl boronicacid PubChem CID: 44119833 IUPAC Name: [3-[ethyl(methyl)carbamoyl]phenyl]boronic acid SMILES: B(C1=CC(=CC=C1)C(=O)N(C)CC)(O)O