missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-hexanol, 98%, Thermo Scientific Chemicals

Product Code. p-7025406
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11405816

Brand: Thermo Scientific Alfa Aesar B20859.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-76-2
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00039616
InChI Key WNDLTOTUHMHNOC-UHFFFAOYSA-N
Synonym 3-ethyl-3-hexanol, 3-hexanol, 3-ethyl, 1,1-diethyl-1-butanol, diethyl n-propyl carbinol, acmc-1ak6h
PubChem CID 69008
IUPAC Name 3-ethylhexan-3-ol
SMILES CCCC(O)(CC)CC

Specifications

Density 0.83
Quantity 1 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3-Ethyl-3-hexanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.