accessibility menu, dialog, popup

UserName

CH3OHH3CH3C

CAS RN 597-76-2

IUPAC Name: 3-ethylhexan-3-ol
Synonyms: 3-ethyl-3-hexanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: WNDLTOTUHMHNOC-UHFFFAOYSA-N
SMILES: CCCC(O)(CC)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results