Learn More
3-Ethoxypropylamine, 99%
Product Code. 11440108
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440108 500 g 500g
11430108 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11440108 Supplier Thermo Scientific Alfa Aesar Supplier No. B23694.36
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 6291-85-6 | C5H13NO | 103.165 g/mol
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.
Solubility
Miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6291-85-6
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008221
InChI Key SOYBEXQHNURCGE-UHFFFAOYSA-N
Synonym 3-ethoxypropylamine, 1-propanamine, 3-ethoxy, 3-ethoxy-1-propanamine, 3-ethoxy-1-propylamine, propylamine, 3-ethoxy, 3-ethoxy-propylamine, 3-aminopropyl ethyl ether, 3-ethyoxypropylamine, 3-ethoxypropyl amine, 3-ethyloxy-propylamine
PubChem CID 22720
IUPAC Name 3-ethoxypropan-1-amine
SMILES CCOCCCN
Specifications
Density 0.86
Boiling Point 136°C to 138°C
Flash Point 32°C (91°F)
Linear Formula CH3CH2O(CH2)3NH2
Refractive Index 1.4185
Quantity 500 g
UN Number UN2733
Beilstein 1732287
Solubility Information Miscible in water.
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 3-Ethoxypropylamine