Learn More
3-Ethoxybenzonitrile, 95%
Product Code. 11481618
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481618 5 g 5g
11491618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11481618 Supplier Thermo Scientific Alfa Aesar Supplier No. A13908.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
CAS: 25117-75-3 | C9H9NO | 147.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25117-75-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001805
InChI Key PFNAUBYHTWDKBO-UHFFFAOYSA-N
Synonym benzonitrile, 3-ethoxy, benzonitrile, m-ethoxy, m-ethoxy benzonitrile, acmc-20an4n, 3-ethoxybenzenecarbonitrile, 3-ethoxybenzoic acid nitrile
PubChem CID 91283
IUPAC Name 3-ethoxybenzonitrile
SMILES CCOC1=CC=CC(=C1)C#N
Specifications
Density 1.047
Boiling Point 241°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 5 g
UN Number UN3276
Formula Weight 147.18
Percent Purity 95%
Chemical Name or Material 3-Ethoxybenzonitrile