Learn More
3-Dodecyldihydrofuran-2,5-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2561-85-5 |
| Molecular Formula | C16H28O3 |
| MDL Number | MFCD00022610 |
| Quantity | 5 g |
| SMILES | O=C(O1)C(CCCCCCCCCCCC)CC1=O |
| Molecular Weight (g/mol) | 268.39 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Dodecyldihydrofuran-2,5-dione |