missing translation for 'onlineSavingsMsg'
Learn More

[3-(Dimethylamino)propyl]triphenylphosphonium Bromide Hydrobromide, TRC

Product Code. 30385963
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385963 10 g
30369752 5 g
30384109 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385963 Supplier Toronto Research Chemicals Supplier No. TRCD49530310G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27710-82-3
Molecular Formula C23H28Br2NP
Molecular Weight (g/mol) 509.3
Synonym Phosphonium, [3-(dimethylamino)propyl]triphenyl-, bromide, hydrobromide (1:1:1) (ACI), Phosphonium, [3-(dimethylamino)propyl]triphenyl-, bromide, hydrobromide (8CI, 9CI), [3-(Dimethylamino)propyl]triphenylphosphonium bromide hydrobromide
SMILES Br.[Br-].CN(C)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C23H27NP.2BrH/c1-24(2)19-12-20-25(21-13-6-3-7-14-21,22-15-8-4-9-16-22)23-17-10-5-11-18-23;;/h3-11,13-18H,12,19-20H2,1-2H3;2*1H/q+1;;/p-1
Formula Weight 507.0326
Chemical Name or Material [3-(Dimethylamino)propyl]triphenylphosphonium Bromide Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.