missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. p-4380092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11966091 1 g 1g
11976091 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966091 Supplier Thermo Scientific Acros Supplier No. 444950010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 325142-87-8
Molecular Formula C14H22BNO2
Molecular Weight (g/mol) 247.14
InChI Key BIJRPONWUBCGES-UHFFFAOYSA-N
Synonym n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-n,n-dimethylamino phenylboronic acid, pinacol ester, 3-n,n-dimethylamino phenylboronic acid pinacol ester, dimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, 3-dimethylamino phenylboronic acid pinacol ester, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, n,n-dimethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
PubChem CID 17998909
IUPAC Name N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N(C)C

Specifications

Melting Point 53°C to 55°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 247.14
Percent Purity 97%
Chemical Name or Material 3-(Dimethylamino)phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-(Dimethylamino)phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.