Learn More
3-Deoxygalactosone (>90%), TRC
Product Code. 30134241
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134241 50 mg 50mg
30136626 5 mg 5mg
30004156 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134241 Supplier Toronto Research Chemicals Supplier No. TRCD23710050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4134-97-8
Molecular Formula C6 H10 O5
Molecular Weight (g/mol) 162.14
Synonym D-threo-Hexos-2-ulose, 3-deoxy-, D-threo-Hexosulose, 3-deoxy- (6CI,7CI,8CI), 3-Deoxy-D-threo-hexos-2-ulose, 3-Deoxy-D-galactosone, 3-Deoxygalactosone, (4R,5R)-4,5,6-Trihydroxy-2-oxohexanal
IUPAC Name (4R,5R)-4,5,6-trihydroxy-2-oxohexanal
SMILES [C@@H](CC(C=O)=O)([C@@H](CO)O)O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H10O5/c7-2-4(9)1-5(10)6(11)3-8/h2,5-6,8,10-11H,1,3H2/t5-,6-/m1/s1
Formula Weight 162.0528
Percent Purity >95%
Chemical Name or Material 3-Deoxy-galactosone
Grade HPLC