missing translation for 'onlineSavingsMsg'
Learn More

3-Demethyl Thiocolchicine, TRC

Product Code. 30132413
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132413 5 mg 5mg
30011598 25 mg 25mg
30006255 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132413 Supplier Toronto Research Chemicals Supplier No. TRCD2313305MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87424-25-7
Molecular Formula C21 H23 N O5 S
Molecular Weight (g/mol) 401.48
Synonym N-[(7S,12aRa)-3-Hydroxy-1,2-dimethoxy-10-(methylsulfanyl)-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide, Acetamide, N-[(7S)-5,6,7,9-tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, Acetamide, N-[5,6,7,9-tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, (S)-, Colchicine, 2-demethylthio- (6CI), Benzo[a]heptalene, acetamide deriv., N-[(7S)-5,6,7,9-Tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]acetamide, 3-Demethylthiocolchicine, 3-Desmethylthiocolchicine, 3-O-Demethylthiocolchicine, NSC 361792
IUPAC Name N-[(7S)-3-hydroxy-1,2-dimethoxy-10-methylsulfanyl-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COc1c(O)cc2CC[C@H](NC(=O)C)C3=CC(=O)C(=CC=C3c2c1OC)SC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H23NO5S/c1-11(23)22-15-7-5-12-9-17(25)20(26-2)21(27-3)19(12)13-6-8-18(28-4)16(24)10-14(13)15/h6,8-10,15,25H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1
Formula Weight 401.1297
Percent Purity >95%
Chemical Name or Material 3-Demethyl Thiocolchicine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.