missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorosalicylic Acid, TRC

Product Code. 30007141
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007141 1 g 1g
30004185 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007141 Supplier Toronto Research Chemicals Supplier No. TRCC3820001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1829-32-9
Molecular Formula C7 H5 Cl O3
Molecular Weight (g/mol) 172.57
IUPAC Name 3-chloro-2-hydroxybenzoic acid
SMILES OC(=O)c1cccc(Cl)c1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5ClO3/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,9H,(H,10,11)
Formula Weight 171.9927
Chemical Name or Material 3-Chlorosalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.