Learn More
3-Chlorocyclohexene, TRC
Product Code. 30361313
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361313 100 mg 100mg
30350398 50 mg -
30354510 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30361313 Supplier Toronto Research Chemicals Supplier No. TRCC384255100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2441-97-6
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.59
Synonym Cyclohexene, 3-chloro- (6CI, 8CI, 9CI, ACI), 3-Chlorocyclohexene (ACI), 1-Chloro-2-cyclohexene, 2-Cyclohexenyl chloride, 3-Chloro-1-cyclohexene, 3-Cyclohexenyl chloride
IUPAC Name 3-chlorocyclohexene
SMILES ClC1CCCC=C1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H9Cl/c7-6-4-2-1-3-5-6/h2,4,6H,1,3,5H2
Formula Weight 116.0393
Chemical Name or Material 3-Chlorocyclohexene