Learn More
3-Chlorobenzal chloride, 95%, Thermo Scientific™
Product Code. 11425097
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425097 50 g 50g
11415097 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11425097 Supplier Thermo Scientific Alfa Aesar Supplier No. B21973.18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15145-69-4
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00082691
InChI Key ACNQJGLSENYFQJ-UHFFFAOYSA-N
PubChem CID 84803
IUPAC Name 1-chloro-3-(dichloromethyl)benzene
SMILES C1=CC(=CC(=C1)Cl)C(Cl)Cl
Tekniske data
Density 1.38
Quantity 50 g
Odor Pungent
Refractive Index 1.566
UN Number UN3265
Synonym 3-chlorobenzal chloride, 1-chloro-3-dichloromethyl benzene, alpha,alpha,3-trichlorotoluene, benzene,1-chloro-3-dichloromethyl, m-chlorbenzalchlorid, 3-chlorobenzalchloride, benzene, 1-chloro-3-dichloromethyl, 3-chlrobenzal chloride, acmc-20an68, 1-dichloromethyl-3-chlorobenzene
Boiling Point 235°C to 237°C
Formula Weight 195.48
Percent Purity 95%
Chemical Name or Material 3-Chlorobenzal chloride
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.