Learn More
3-chloro-5-fluoro-2-hydroxybenzoic Acid, TRC
Product Code. 30367609
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367609 50 mg 50mg
30352909 100 mg 100mg
30364206 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367609 Supplier Toronto Research Chemicals Supplier No. TRCC37505850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4068-62-6
Molecular Formula C7H4O3FCl
Molecular Weight (g/mol) 190.56
IUPAC Name 3-chloro-5-fluoro-2-hydroxybenzoic acid
SMILES OC(=O)c1cc(F)cc(Cl)c1O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7H4ClFO3/c8-5-2-3(9)1-4(6(5)10)7(11)12/h1-2,10H,(H,11,12)
Formula Weight 189.9833
Chemical Name or Material 3-chloro-5-fluoro-2-hydroxybenzoic Acid