Learn More
3-Chloro-4-iodoaniline, 98%, Thermo Scientific™
Product Code. 11492387
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11492387
Brand: Thermo Scientific Alfa Aesar A14256.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chloro-4-iodoaniline used as a intermediate for organic syntheses.
Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135050-44-1
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00051845
InChI Key ONZHMGRKWJMTDE-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-iodo, 3-chloro-4-iodobenzenamine, 4-amino-2-chloroiodobenzene, 3-chloro-4-iodophenylamine, pubchem3812, 3-chloro4-iodoaniline, zlchem 1106, acmc-209bxp, 3-chloro-4-iodo-aniline, 3-chloro-4-iodo aniline
PubChem CID 282926
IUPAC Name 3-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C(Cl)=C1
Specifications
Melting Point 65°C to 71°C
Quantity 5 g
UN Number UN2811
Beilstein 3236464
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-iodoaniline