Learn More
3-Chloro-4-fluorobenzonitrile, 98+%
Product Code. 11448576
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448576 5 g 5g
11458576 25 g 25g
11468576 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11448576 Supplier Thermo Scientific Alfa Aesar Supplier No. A15987.06
Request Bulk or Custom Format
CAS: 117482-84-5 | C7H3ClFN | 155.556 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 117482-84-5
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00015431
InChI Key VAHXXQJJZKBZDX-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-benzonitrile, benzonitrile, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenecarbonitrile, pubchem1561, acmc-1bnmf, 3-chloro-4-florobenzonitrile, 3-chloro-4-fluorobenzontrile, ksc174m4n, 3-chloro,4-fluorobenzonitrile, 3-chloro4-fluorobenzo-nitrile
PubChem CID 145525
IUPAC Name 3-chloro-4-fluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)F
Specifications
Melting Point 68°C to 72°C
Quantity 5 g
UN Number UN3439
Beilstein 4740501
Formula Weight 155.56
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-fluorobenzonitrile